Anti-PI 4 Kinase Type 2 beta Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
P4186-28N-FITC
USBIP4186-28N-FITCEA 884 EUR
USBIP4186-28N-FITC
Anti-PI 4 Kinase Type 2 beta Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: PI 4 Kinase Type 2 beta
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR