Anti-Pituitary Growth/Lactation Coupled Transporter Rabbit Polyclonal Antibody
Lieferant: US Biological
P4212
USBIP4212EA 696 EUR
USBIP4212
Anti-Pituitary Growth/Lactation Coupled Transporter Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:200
Type: Primary
Antigen: Pituitary Growth/Lactation Coupled Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human