Anti-Platelet Derived Growth Factor BB Rabbit Polyclonal Antibody

Lieferant: US Biological
P4258-17G
USBIP4258-17GEA 560 EUR
USBIP4258-17G
Anti-Platelet Derived Growth Factor BB Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000-1:8000

Type: Primary
Antigen: Platelet Derived Growth Factor BB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR