Anti-Propionyl Coenzyme A Carboxylase beta Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: US Biological
P9052-06B-APC
USBIP9052-06B-APCEA 884 EUR
USBIP9052-06B-APC
Anti-Propionyl Coenzyme A Carboxylase beta Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: Propionyl Coenzyme A Carboxylase beta
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR