Anti-Propionyl Coenzyme A Carboxylase beta Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
P9052-06B-BIOT
USBIP9052-06B-BIOTEA 884 EUR
USBIP9052-06B-BIOT
Anti-Propionyl Coenzyme A Carboxylase beta Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Propionyl Coenzyme A Carboxylase beta
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR