Anti-PCSK9 Goat Polyclonal Antibody

Lieferant: US Biological
P9052-40E
USBIP9052-40EEA 601 EUR
USBIP9052-40E
Anti-PCSK9 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0.1-0.5 ?g/ml

Type: Primary
Antigen: PCSK9 (proprotein convertase subtilisin/kexin type 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR