Anti-Prostaglandin D2 Synthase Rabbit Polyclonal Antibody

Lieferant: US Biological
P9053-24B
USBIP9053-24BEA 614 EUR
USBIP9053-24B
Anti-Prostaglandin D2 Synthase Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: Prostaglandin D2 Synthase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR