Anti-Prostaglandin E1 Rabbit Polyclonal Antibody
Lieferant: US Biological
P9053-28C
USBIP9053-28CEA 489 EUR
USBIP9053-28C
Anti-Prostaglandin E1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prostaglandin E1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen