Anti-Prostaglandin E2 Rabbit Polyclonal Antibody

Lieferant: US Biological
P9053-28E
USBIP9053-28EEA 389 EUR
USBIP9053-28E
Anti-Prostaglandin E2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prostaglandin E2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR