Anti-Prostaglandin F2 alpha, 8-iso- Rabbit Polyclonal Antibody

Lieferant: US Biological
P9053-34K
USBIP9053-34KEA 795 EUR
USBIP9053-34K
Anti-Prostaglandin F2 alpha, 8-iso- Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Prostaglandin F2 alpha, 8-iso-
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR