Anti-Prostatic Acid Phosphatase Mouse Monoclonal Antibody [clone: 9E102]

Lieferant: US Biological
P9054-67B-100
USBIP9054-67B-100EA 748 EUR
USBIP9054-67B-100
Anti-Prostatic Acid Phosphatase Mouse Monoclonal Antibody [clone: 9E102]
Antikörper
Recommended dilution: ELISA: 1:200-1:400

Type: Primary
Antigen: Prostatic Acid Phosphatase
Clonality: monoclonal
Clone: 9E+102
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR