Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
Lieferant: US Biological
P9102-70F
USBIP9102-70FEA 599 EUR
USBIP9102-70F
Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Immunochemistry: 1:200-1:800
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Cat, Dog, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat, Sheep