Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
Lieferant: US Biological
P9102-70M
USBIP9102-70MEA 566 EUR
USBIP9102-70M
Anti-Protein Gene Product 9.5 Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 0.1-1 μg/ml
Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey