Anti-PRKCD Rabbit Polyclonal Antibody

Lieferant: US Biological
P9103-19S
USBIP9103-19SEA 809 EUR
USBIP9103-19S
Anti-PRKCD Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PRKCD (protein kinase C, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR