Anti-PRKCQ Rabbit Polyclonal Antibody

Lieferant: US Biological
P9103-68C
USBIP9103-68CEA 380 EUR
USBIP9103-68C
Anti-PRKCQ Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PRKCQ (protein kinase C, theta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR