Anti-PP1 gamma1 Rabbit Polyclonal Antibody

Lieferant: US Biological
P9107-20A
USBIP9107-20AEA 736 EUR
USBIP9107-20A
Anti-PP1 gamma1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PP1 gamma1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR