Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT18.7]

Lieferant: US Biological
P9181-90R
USBIP9181-90REA 568 EUR
USBIP9181-90R
Anti-PINK1 Mouse Monoclonal Antibody [clone: 38CT18.7]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: monoclonal
Clone: 38CT18.7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR