Anti-RAD51 Mouse Monoclonal Antibody [clone: 6A174]

Lieferant: US Biological
R0451-01E
USBIR0451-01EEA 695 EUR
USBIR0451-01E
Anti-RAD51 Mouse Monoclonal Antibody [clone: 6A174]
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml; Immunoprecipitation: 10 ?g/Immunoprecipitation: Reaction

Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: monoclonal
Clone: 6A174
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR