Anti-RAD51 Mouse Monoclonal Antibody [clone: 13E4]

Lieferant: US Biological
R0451-01G1
USBIR0451-01G1EA 755 EUR
USBIR0451-01G1
Anti-RAD51 Mouse Monoclonal Antibody [clone: 13E4]
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: monoclonal
Clone: 130000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR