Anti-RAD51 Rabbit Polyclonal Antibody
Lieferant: US Biological
R0451-01J
USBIR0451-01JEA 542 EUR
USBIR0451-01J
Anti-RAD51 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1-0.5 ?g/ml; Immunochemistry: 0.5-1 ?g/ml.
Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat