Anti-RAD51 Rabbit Polyclonal Antibody

Lieferant: US Biological
R0451-01K
USBIR0451-01KEA 542 EUR
USBIR0451-01K
Anti-RAD51 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: RAD51 (RAD51 recombinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR