Anti-Renin, Pro/Mature Mouse Monoclonal Antibody [clone: 7H238]

Lieferant: US Biological
R1460-05
USBIR1460-05EA 871 EUR
USBIR1460-05
Anti-Renin, Pro/Mature Mouse Monoclonal Antibody [clone: 7H238]
Antikörper
Recommended dilution: Immunoprecipitation: 25 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Renin, Pro/Mature
Clonality: monoclonal
Clone: 7H238
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR