Anti-Renin-1 Goat Polyclonal Antibody

Lieferant: US Biological
R1460-40
USBIR1460-40EA 897 EUR
USBIR1460-40
Anti-Renin-1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1-0.2?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Renin-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR