Anti-Renin-1 Goat Polyclonal Antibody (Biotin)
Lieferant: US Biological
R1460-40A
USBIR1460-40AEA 944 EUR
USBIR1460-40A
Anti-Renin-1 Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.1-0.4?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Renin-1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse