Anti-Renin R Goat Polyclonal Antibody

Lieferant: US Biological
R1475-10
USBIR1475-10EA 871 EUR
USBIR1475-10
Anti-Renin R Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1.0 ?g/ml

Type: Primary
Antigen: Renin R
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR