Anti-Respiratory Syncytial Virus, pan Goat Polyclonal Antibody

Lieferant: US Biological
R1595-30A
USBIR1595-30AEA 546 EUR
USBIR1595-30A
Anti-Respiratory Syncytial Virus, pan Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Respiratory Syncytial Virus, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR