Anti-RB1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
R1660-05G3BIOT
USBIR1660-05G3BIOTEA 884 EUR
USBIR1660-05G3BIOT
Anti-RB1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR