Anti-ZMIZ1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
R1690-17-BIOT
USBIR1690-17-BIOTEA 884 EUR
USBIR1690-17-BIOT
Anti-ZMIZ1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ZMIZ1 (zinc finger, MIZ-type containing 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR