Anti-Retinoic Acid Receptor alpha Rabbit Polyclonal Antibody

Lieferant: US Biological
R1691-01A
USBIR1691-01AEA 477 EUR
USBIR1691-01A
Anti-Retinoic Acid Receptor alpha Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Retinoic Acid Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR