Anti-Rhodamine Mouse Monoclonal Antibody [clone: NI 308]

Lieferant: US Biological
R1998-12F
USBIR1998-12FEA 641 EUR
USBIR1998-12F
Anti-Rhodamine Mouse Monoclonal Antibody [clone: NI 308]
Antikörper
Type: Tag
Antigen: Rhodamine
Clonality: monoclonal
Clone: NI 308
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR