Anti-Rho Guanine Nucleotide Exchange Factor H1 Rabbit Monoclonal Antibody [clone: 7H57]

Lieferant: US Biological
R1998-26
USBIR1998-26EA 608 EUR
USBIR1998-26
Anti-Rho Guanine Nucleotide Exchange Factor H1 Rabbit Monoclonal Antibody [clone: 7H57]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: Rho Guanine Nucleotide Exchange Factor H1
Clonality: monoclonal
Clone: 7H57
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR