Anti-Rho guanine nucleotide exchange factor 4 Goat Polyclonal Antibody

Lieferant: US Biological
R1998-27B
USBIR1998-27BEA 809 EUR
USBIR1998-27B
Anti-Rho guanine nucleotide exchange factor 4 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 2-4?g/ml

Type: Primary
Antigen: Rho guanine nucleotide exchange factor 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR