Anti-RRM2 Mouse Monoclonal Antibody [clone: 1E1]

Lieferant: US Biological
R2029-11B
USBIR2029-11BEA 703 EUR
USBIR2029-11B
Anti-RRM2 Mouse Monoclonal Antibody [clone: 1E1]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: RRM2 (ribonucleotide reductase M2)
Clonality: monoclonal
Clone: 10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR