Anti-Receptor-Interacting Serine/Threonine-Protein Kinase Mouse Monoclonal Antibody [clone: 9F638]

Lieferant: US Biological
R2031-73N
USBIR2031-73NEA 574 EUR
USBIR2031-73N
Anti-Receptor-Interacting Serine/Threonine-Protein Kinase Mouse Monoclonal Antibody [clone: 9F638]
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 1:12000

Type: Primary
Antigen: Receptor-Interacting Serine/Threonine-Protein Kinase
Clonality: monoclonal
Clone: 9F638
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR