Anti-RLA, DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K114]
Lieferant: US Biological
R2032-50A
USBIR2032-50AEA 607 EUR
USBIR2032-50A
Anti-RLA, DR Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K114]
Antikörper
Type: Primary
Antigen: RLA, DR
Clonality: monoclonal
Clone: 5K114
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Rabbit