Anti-ROK alpha/ROCK-II Rabbit Monoclonal Antibody [clone: 9H271]

Lieferant: US Biological
R2998-26C
USBIR2998-26CEA 736 EUR
USBIR2998-26C
Anti-ROK alpha/ROCK-II Rabbit Monoclonal Antibody [clone: 9H271]
Antikörper
Recommended dilution: Western Blot: 1:500-1:5000

Type: Primary
Antigen: ROK alpha/ROCK-II
Clonality: monoclonal
Clone: 9H271
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR