Anti-S1 Rabbit Polyclonal Antibody

Lieferant: US Biological
S0015-30
USBIS0015-30EA 374 EUR
USBIS0015-30
Anti-S1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml

Type: Tag
Antigen: S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR