Anti-S1 Nuclease Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S0050-02B
USBIS0050-02BEA 542 EUR
USBIS0050-02B
Anti-S1 Nuclease Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: S1 Nuclease
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR