Anti-S-Adenosyl-Homocysteine Rabbit Polyclonal Antibody

Lieferant: US Biological
S0056-21
USBIS0056-21EA 761 EUR
USBIS0056-21
Anti-S-Adenosyl-Homocysteine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S-Adenosyl-Homocysteine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR