Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 10F420]
Lieferant: US Biological
S0060-23A
USBIS0060-23AEA 796 EUR
USBIS0060-23A
Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 10F420]
Antikörper
Recommended dilution: ELISA: 1:20-1:200
Type: Primary
Antigen: Salmonella sp.
Clonality: Monoclonal
Clone: 10F420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen