Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 42-77]

Lieferant: US Biological
S0060-24E
USBIS0060-24EEA 414 EUR
USBIS0060-24E
Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 42-77]
Antikörper
Recommended dilution: ELISA: 1:10

Type: Primary
Antigen: Salmonella sp.
Clonality: Monoclonal
Clone: 42-77
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR