Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 10F416]

Lieferant: US Biological
S0060-26H
USBIS0060-26HEA 575 EUR
USBIS0060-26H
Anti-Salmonella sp. Mouse Monoclonal Antibody [clone: 10F416]
Antikörper
Type: Primary
Antigen: Salmonella sp.
Clonality: Monoclonal
Clone: 10F416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR