Anti-Salmonella sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5E433]

Lieferant: US Biological
S0060-53A
USBIS0060-53AEA 822 EUR
USBIS0060-53A
Anti-Salmonella sp. Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5E433]
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50; FLISA: 1:20-1:200

Type: Primary
Antigen: Salmonella sp.
Clonality: Monoclonal
Clone: 5E433
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR