Anti-SARM1 Rabbit Polyclonal Antibody
Lieferant: US Biological
S0097-70
USBIS0097-70EA 647 EUR
USBIS0097-70
Anti-SARM1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5 to 2?g/ml
Type: Primary
Antigen: SARM1 (sterile alpha and TIR motif containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse