Anti-Semaphorin 5A Rabbit Polyclonal Antibody

Lieferant: US Biological
S0668-57B
USBIS0668-57BEA 649 EUR
USBIS0668-57B
Anti-Semaphorin 5A Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR