Anti-Semaphorin 5A Sheep Polyclonal Antibody

Lieferant: US Biological
S0668-57E
USBIS0668-57EEA 857 EUR
USBIS0668-57E
Anti-Semaphorin 5A Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR