Anti-Semaphorin 5A Goat Polyclonal Antibody

Lieferant: US Biological
S0668-57G
USBIS0668-57GEA 601 EUR
USBIS0668-57G
Anti-Semaphorin 5A Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 03?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: Semaphorin 5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR