Anti-Serotonin Transporter Rabbit Polyclonal Antibody

Lieferant: US Biological
S1001-25B
USBIS1001-25BEA 843 EUR
USBIS1001-25B
Anti-Serotonin Transporter Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:800-1:1000

Type: Primary
Antigen: Serotonin Transporter
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR