Anti-SF-20 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S1012-83M1-25
USBIS1012-83M1-25EA 324 EUR
USBIS1012-83M1-25
Anti-SF-20 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: SF-20
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR