Anti-SF-20 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
S1012-83P
USBIS1012-83PEA 663 EUR
USBIS1012-83P
Anti-SF-20 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.25-1 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: SF-20
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR